Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036893.2 | NP_001031970.1 | 5 - 6 | 1259 | 92 | |
NM_001344125.1 | NP_001332507.1 | 5 - 6 | 737 | 92 | ||
NM_122989.4 | NP_198447.2 | 5 - 6 | 1283 | 92 | ||
GenBank mRNA | AK230350.1 | - | 5 - 6 | 1201 | 92 | |
AY093090.1 | - | 5 - 6 | 1259 | 92 | ||
BT010549.1 | - | 5 - 6 | 692 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |