Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344271.1 | NP_001330505.1 | 5 - 6 | 840 | 80 | |
NM_123247.5 | NP_198702.2 | 5 - 6 | 968 | 80 | ||
GenBank mRNA | AJ630499.1 | - | 5 - 6 | 786 | 80 | |
AY568671.1 | - | 5 - 6 | 786 | 80 | ||
BT033131.1 | - | 5 - 6 | 786 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |