Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc2cAt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123304.4 | NP_198758.2 | 1 - 2 | 624 | 70 | |
GenBank mRNA | BT046183.1 | - | 1 - 2 | 323 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |