Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203515.1 | NP_001190444.1 | 2 - 3 | 954 | 78 | |
NM_001344363.1 | NP_001331463.1 | 2 - 3 | 954 | 78 | ||
NM_123450.5 | NP_198901.1 | 2 - 3 | 1407 | 78 | ||
GenBank mRNA | AF462833.1 | - | 2 - 3 | 793 | 78 | |
AY058093.1 | - | 2 - 3 | 789 | 78 | ||
AY079029.1 | - | 2 - 3 | 793 | 78 | ||
AY142030.1 | - | 1 - 2 | 533 | 78 | ||
L47479.1 | - | 2 - 3 | 647 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |