Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123464.2 | NP_198915.1 | 2 - 3 | 475 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |