Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036914.1 | NP_001031991.1 | 2 - 3 | 1122 | 58 | |
NM_001344387.1 | NP_001330263.1 | 2 - 3 | 1588 | 58 | ||
NM_001344388.1 | NP_001330264.1 | 2 - 3 | 1122 | 58 | ||
NM_123480.4 | NP_198931.1 | 2 - 3 | 1588 | 58 | ||
GenBank mRNA | AF089003.1 | - | 2 - 3 | 1090 | 58 | |
AF160500.1 | - | 2 - 3 | 1184 | 58 | ||
AY140076.1 | - | 2 - 3 | 1122 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |