Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123592.4 | NP_568604.1 | 4 - 5 | 2078 | 68 | |
GenBank mRNA | AY126987.1 | - | 4 - 5 | 1993 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |