Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123646.4 | NP_568613.1 | 1 - 2 | 480 | 72 | |
GenBank mRNA | AY059898.1 | - | 1 - 2 | 355 | 72 | |
AY093362.1 | - | 1 - 2 | 109 | 72 | ||
DQ122931.1 | - | 1 - 2 | 109 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |