Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TZP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344491.1 | NP_001330182.1 | 3 - 4 | 2740 | 86 | |
NM_001344492.1 | NP_001330183.1 | 4 - 5 | 2569 | 86 | ||
NM_001344493.1 | NP_001330181.1 | 4 - 5 | 2275 | 86 | ||
NM_123729.3 | NP_199176.1 | 4 - 5 | 2290 | 86 | ||
GenBank mRNA | BT003987.1 | - | 4 - 5 | 2268 | 86 | |
BT005699.1 | - | 3 - 4 | 2139 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |