Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123745.6 | NP_199191.1 | 1 - 2 | 774 | 77 | |
GenBank mRNA | AJ012586.1 | - | 1 - 2 | 545 | 77 | |
AY072193.1 | - | 1 - 2 | 529 | 77 | ||
AY117323.1 | - | 1 - 2 | 517 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |