Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203535.1 | NP_001190464.1 | 18 - 19 | 3141 | 140 | |
NM_001344505.1 | NP_001331600.1 | 17 - 18 | 3064 | 140 | ||
NM_001344506.1 | NP_001331599.1 | 17 - 18 | 2881 | 140 | ||
NM_123748.3 | NP_199194.1 | 17 - 18 | 2936 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |