Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123875.3 | NP_199320.1 | 2 - 3 | 835 | 75 | |
GenBank mRNA | BT010864.1 | - | 2 - 3 | 696 | 75 | |
BT012618.1 | - | 2 - 3 | 674 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |