Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344590.1 | NP_001332060.1 | 29 - 30 | 2734 | 65 | |
NM_123882.5 | NP_199327.4 | 29 - 30 | 2652 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |