Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036937.1 | NP_001032014.1 | 8 - 9 | 1847 | 71 | |
NM_001344606.1 | NP_001330613.1 | 8 - 9 | 1936 | 71 | ||
NM_123898.3 | NP_199343.1 | 8 - 9 | 1851 | 71 | ||
GenBank mRNA | AK117140.1 | - | 8 - 9 | 1848 | 71 | |
BT006482.1 | - | 8 - 9 | 1825 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |