Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123906.5 | NP_199351.2 | 1 - 2 | 570 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |