Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G45430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203544.2 | NP_001190473.1 | 11 - 12 | 1823 | 77 | |
NM_123911.2 | NP_568646.1 | 11 - 12 | 1585 | 77 | ||
GenBank mRNA | BT001210.1 | - | 8 - 9 | 716 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |