Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G45430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203544.2 | NP_001190473.1 | 5 - 6 | 1266 | 128 | |
NM_123911.2 | NP_568646.1 | 5 - 6 | 1028 | 128 | ||
GenBank mRNA | BT001210.1 | - | 2 - 3 | 159 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |