Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123965.2 | NP_199409.1 | 3 - 4 | 520 | 87 | |
GenBank mRNA | U43026.1 | - | 3 - 4 | 326 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |