Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123993.3 | NP_199436.1 | 5 - 6 | 1774 | 71 | |
GenBank mRNA | AK176505.1 | - | 5 - 6 | 974 | 71 | |
AK220637.1 | - | 5 - 6 | 974 | 71 | ||
BT046198.1 | - | 5 - 6 | 929 | 71 | ||
M86990.1 | - | 5 - 6 | 948 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |