Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124024.2 | NP_199466.1 | 10 - 11 | 2947 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |