Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124050.2 | NP_568669.1 | 1 - 2 | 96 | 82 | |
GenBank mRNA | DQ447047.1 | - | 1 - 2 | 67 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |