Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124205.4 | NP_199641.1 | 6 - 7 | 1413 | 105 | |
GenBank mRNA | AY049265.1 | - | 6 - 7 | 1389 | 105 | |
AY065428.1 | - | 6 - 7 | 1355 | 105 | ||
AY090283.1 | - | 6 - 7 | 1325 | 105 | ||
AY096379.1 | - | 6 - 7 | 1325 | 105 | ||
U72351.1 | - | 6 - 7 | 1360 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |