Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124224.3 | NP_568699.1 | 1 | 482 | 63 | |
GenBank mRNA | AY062857.1 | - | 1 | 319 | 63 | |
BT006510.1 | - | 1 | 267 | 63 |
Gene Symbol: AT5G48480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124223.4 | NP_568698.1 | 2 | 1477 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |