Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344853.1 | NP_001331052.1 | 1 - 2 | 379 | 73 | |
NM_001344854.1 | NP_001331053.1 | 1 - 2 | 332 | 73 | ||
NM_124352.4 | NP_199785.2 | 1 - 2 | 379 | 73 | ||
GenBank mRNA | AK227087.1 | - | 1 - 2 | 273 | 73 | |
AY912280.1 | - | 1 - 2 | 230 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |