Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124359.4 | NP_199792.1 | 1 - 2 | 346 | 167 | |
GenBank mRNA | AY094459.1 | - | 1 - 2 | 296 | 167 | |
BT002664.1 | - | 1 - 2 | 221 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons |