Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124377.3 | NP_568718.1 | 1 - 2 | 743 | 93 | |
GenBank mRNA | AY056431.1 | - | 1 - 2 | 738 | 93 | |
AY139774.1 | - | 1 - 2 | 738 | 93 | ||
BT004536.1 | - | 1 - 2 | 603 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |