Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344917.1 | NP_001330684.1 | 2 - 3 | 454 | 61 | |
NM_001344918.1 | NP_001330683.1 | 3 - 4 | 252 | 61 | ||
NM_001344919.1 | NP_001330687.1 | 3 - 4 | 252 | 61 | ||
NM_001344920.1 | NP_001330686.1 | 3 - 4 | 258 | 61 | ||
NM_001344921.1 | NP_001330685.1 | 1 - 2 | 152 | 61 | ||
NM_124489.3 | NP_199923.1 | 3 - 4 | 290 | 61 | ||
GenBank mRNA | AY085077.1 | - | 3 - 4 | 223 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |