Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124527.5 | NP_199961.1 | 1 - 2 | 362 | 75 | |
GenBank mRNA | BT004129.1 | - | 1 - 2 | 322 | 75 | |
BT004990.1 | - | 1 - 2 | 133 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |