Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125943.1 | NP_001119415.1 | 1 - 2 | 342 | 80 | |
NM_124550.2 | NP_568763.1 | 1 - 2 | 473 | 80 | ||
GenBank mRNA | AY070064.1 | - | 1 - 2 | 288 | 80 | |
AY084820.1 | - | 1 - 2 | 341 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |