Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124589.3 | NP_200023.1 | 2 - 3 | 384 | 76 | |
GenBank mRNA | BT030092.1 | - | 2 - 3 | 229 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |