Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124635.4 | NP_568775.1 | 9 - 10 | 1135 | 105 | |
GenBank mRNA | AF360236.1 | - | 9 - 10 | 998 | 105 | |
AY040035.1 | - | 8 - 9 | 812 | 105 | ||
DQ324529.1 | - | 8 - 9 | 812 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |