Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BETA-OHASE 2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124636.4 | NP_200070.1 | 4 - 5 | 877 | 112 | |
GenBank mRNA | AY074394.1 | - | 4 - 5 | 660 | 112 | |
AY117225.1 | - | 4 - 5 | 638 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |