Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124661.3 | NP_568777.1 | 2 - 3 | 777 | 151 | |
GenBank mRNA | AB493790.1 | - | 2 - 3 | 550 | 151 | |
AF418310.1 | - | 2 - 3 | 550 | 151 | ||
AK230436.1 | - | 2 - 3 | 694 | 151 | ||
BT026451.1 | - | 2 - 3 | 550 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |