Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124696.4 | NP_568787.1 | 2 - 3 | 864 | 87 | |
GenBank mRNA | AY091086.1 | - | 2 - 3 | 792 | 87 | |
AY123027.1 | - | 2 - 3 | 763 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |