Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMEPCRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345052.1 | NP_001332265.1 | 1 - 2 | 1249 | 93 | |
NM_124716.3 | NP_200149.1 | 1 - 2 | 1162 | 93 | ||
GenBank mRNA | AF360340.1 | - | 1 - 2 | 1106 | 93 | |
AY051077.1 | - | 1 - 2 | 1066 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |