Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336558.1 | NP_001323747.1 | 4 - 5 | 2016 | 154 | |
NM_129103.5 | NP_565806.1 | 4 - 5 | 1876 | 154 | ||
GenBank mRNA | AY150402.1 | - | 2 - 3 | 1354 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |