Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336567.1 | NP_001325149.1 | 23 - 24 | 2938 | 117 | |
NM_129117.5 | NP_565811.2 | 23 - 24 | 2632 | 117 | ||
GenBank mRNA | AF367246.1 | - | 23 - 24 | 2378 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |