Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129117.5 | NP_565811.2 | 3 - 4 | 649 | 77 | |
GenBank mRNA | AF367246.1 | - | 3 - 4 | 395 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Arabidopsis thaliana Probe spans exons |