Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTF2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036412.1 | NP_001031489.1 | 4 - 5 | 906 | 157 | |
NM_129121.3 | NP_565814.1 | 5 - 6 | 864 | 157 | ||
NM_201877.3 | NP_973606.1 | 5 - 6 | 863 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |