Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129124.4 | NP_181112.1 | 7 - 8 | 1401 | 100 | |
GenBank mRNA | AY074357.1 | - | 7 - 8 | 1373 | 100 | |
AY096691.1 | - | 7 - 8 | 1165 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |