Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336578.1 | NP_001318361.1 | 4 - 5 | 1492 | 95 | |
NM_129148.2 | NP_181133.1 | 4 - 5 | 1492 | 95 | ||
GenBank mRNA | DQ446601.1 | - | 4 - 5 | 1269 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |