Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129229.4 | NP_181212.2 | 11 - 12 | 1154 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |