Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336608.1 | NP_001324358.1 | 10 - 11 | 1004 | 98 | |
NM_129229.4 | NP_181212.2 | 10 - 11 | 1081 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |