Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129289.5 | NP_181270.1 | 2 - 3 | 568 | 71 | |
GenBank mRNA | AY072359.1 | - | 2 - 3 | 561 | 71 | |
AY114725.1 | - | 2 - 3 | 509 | 71 | ||
AY687991.1 | - | 2 - 3 | 509 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |