Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129364.4 | NP_565881.1 | 1 - 2 | 391 | 73 | |
GenBank mRNA | AY052669.1 | - | 1 - 2 | 111 | 73 | |
AY063730.1 | - | 1 - 2 | 95 | 73 | ||
AY065187.1 | - | 1 - 2 | 177 | 73 | ||
AY114636.1 | - | 1 - 2 | 95 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |