Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129478.5 | NP_565902.1 | 5 - 6 | 740 | 82 | |
GenBank mRNA | AF369573.1 | - | 5 - 6 | 610 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |