Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125006.1 | NP_001118478.1 | 3 - 4 | 320 | 80 | |
NM_129536.5 | NP_030524.1 | 3 - 4 | 425 | 80 | ||
GenBank mRNA | BT025792.1 | - | 3 - 4 | 286 | 80 | |
M98336.1 | - | 3 - 4 | 301 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |