Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129567.4 | NP_565923.1 | 2 - 3 | 516 | 119 | |
GenBank mRNA | AK175658.1 | - | 2 - 3 | 346 | 119 | |
AK176037.1 | - | 2 - 3 | 346 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |