Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129588.5 | NP_181559.1 | 1 - 2 | 402 | 93 | |
GenBank mRNA | AJ312191.1 | - | 1 - 2 | 364 | 93 | |
AY062795.1 | - | 1 - 2 | 388 | 93 | ||
AY081615.1 | - | 1 - 2 | 322 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |