Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129617.5 | NP_181587.2 | 1 - 2 | 480 | 67 | |
GenBank mRNA | BT022032.1 | - | 1 - 2 | 189 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |